missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver Nitrate, 0.045 Normal, Ricca Chemical

Chemical Identifiers

CAS 7732-18-5,7761-88-8
CAS Min % 97.0,0.74
CAS Max % 100.0,0.77
Physical Form Liquid
Chemical Name or Material Silver Nitrate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
R696500010F Ricca Chemical Company
RIR696500010F
Cubitainer 10L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7761-88-8
100.0,0.77
Silver Nitrate
97.0,0.74
Liquid

Specifications

Liquid
10L
0.045Normal
7761-88-8,7732-18-5
Silver Nitrate
Clear colorless
0.77,100.0
97.0,0.74
Cubitainer