Please login to your online account to display your discounted pricing

Silver Nitrate, 0.0282N, 1mL = 1mg Cl, for Chloride Titrations, Ricca Chemical

$87.79 - $209.55

Chemical Identifiers

Chemical Name or Material Silver Nitrate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

6950-1

 
ricca chemical company
RI69501
Amber Plastic 4L Each for $209.55

6950-32

  SDS
ricca chemical company
RI695032
Amber Plastic 1L Each for $87.79
Description & Specifications

Specifications

Quantity 4L
Equivalence 1mL = 1mg Cl
Chemical Name or Material Silver Nitrate
Concentration or Composition (by Analyte or Components) 0.0282N
Packaging Amber Plastic

For Chloride Titrations