missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
For Chloride Titrations
For Chloride Titrations
Supplier: Ricca Chemical Company 695016
Specifications
Silver Nitrate | |
7761-88-8,7732-18-5 | |
100.0,0.49 | |
MFCD00003414 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
silver(1+) nitrate | |
24470 | |
Amber Poly Bottle | |
500 mL | |
0.0282Normal |
Colorless | |
98.0,0.47 | |
AgNO3 | |
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
[Ag+].[O-][N+]([O-])=O | |
169.87 | |
CHEBI:32130 | |
1 mL = 1 mg Cl | |
Liquid |
Chemical Identifiers
7761-88-8 | |
169.87 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
24470 | |
silver(1+) nitrate |
AgNO3 | |
MFCD00003414 | |
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
CHEBI:32130 | |
[Ag+].[O-][N+]([O-])=O |