Please login to your online account to display your discounted pricing

Silver Nitrate, 0.0200 Normal, Ricca Chemical

$92.33 - $92.33

Chemical Identifiers

Chemical Name or Material Silver Nitrate
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 7761-88-8
CAS Min % 0.33
CAS Max % 0.35
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

6915-32

 
ricca chemical company
RI691532
Amber Poly 1L Each for $92.33
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 1L
Concentration or Composition Notes 0.0200Normal
CAS Min % 98.0
CAS Max % 100.0
CAS 7732-18-5
Chemical Name or Material Silver Nitrate
CAS 7761-88-8
Packaging Amber Poly
CAS Min % 0.33
CAS Max % 0.35