missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Silver Nitrate, 0.0025 N in Methanol, Ricca Chemical
Chemical Identifiers
CAS |
7761-88-8 |
Molecular Formula |
AgNO3 |
Molecular Weight (g/mol) |
169.87 |
MDL Number |
MFCD00003414 |
InChI Key |
SQGYOTSLMSWVJD-UHFFFAOYSA-N |
Synonym |
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol |
PubChem CID |
24470 |
ChEBI |
CHEBI:32130 |
IUPAC Name |
silver(1+) nitrate |
SMILES |
[Ag+].[O-][N+]([O-])=O |
View More Specs
Products
1
Description
Specifications
Chemical Identifiers
|
7761-88-8 |
|
169.87 |
|
SQGYOTSLMSWVJD-UHFFFAOYSA-N |
|
24470 |
|
silver(1+) nitrate |
|
AgNO3 |
|
MFCD00003414 |
|
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol |
|
CHEBI:32130 |
|
[Ag+].[O-][N+]([O-])=O |
Specifications
|
Colorless |
|
0.05,98.0,0.31 |
|
AgNO3 |
|
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol |
|
[Ag+].[O-][N+]([O-])=O |
|
169.87 |
|
CHEBI:32130 |
|
Methanol |
|
Liquid |
|
Silver Nitrate |
|
7761-88-8,67-56-1,7732-18-5 |
|
0.05,100.0,0.32 |
|
MFCD00003414 |
|
SQGYOTSLMSWVJD-UHFFFAOYSA-N |
|
silver(1+) nitrate |
|
24470 |
|
Amber Poly Bottle |
|
1 L |
|
0.0025 N |
SDS
missing translation for 'documents'