missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver nanopowder, APS 20-40nm, 99.9% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04550906

 View more versions of this product

Catalog No. AA4550906



Description

Description

Silver nanopowder is involved in the coating of silver and is used for medicinal purposes and in medical devices. Its larger surface area to volume ratio and greater activity of nano-silver are utilized in modern biochemical applications and drug delivery. It is also used in the fabrication of graphite epoxy nanocomposites and optical fiber sensors. Furthermore, it is involved in the formulation of water based ink.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Silver nanopowder
7440-22-4
Ag
UN3077
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
23954
99.9%
APS 20 to 40nm
No Oxide layer
(metals basis)
MFCD00003397
5 g
Soluble in nitric acid.
[Ag]
107.87
CHEBI:9141
Characteristic
Nanopowder

Chemical Identifiers

7440-22-4
107.87
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
CHEBI:9141
[Ag]
Safety and Handling

Safety and Handling

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 232-033-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only