missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silver nanoparticles, 50nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 425nm absorption, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J67065AC
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Yellow | |
0.02mg/mL, supplied in 2mM sodium citrate, 425nm absorption | |
Ag | |
25 mL | |
BQCADISMDOOEFD-UHFFFAOYSA-N | |
silver | |
23954 | |
Odorless | |
Light sensitive |
Silver nanoparticles | |
7440-22-4 | |
MFCD00003397 | |
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber | |
[Ag] | |
107.87 | |
CHEBI:9141 | |
50nm | |
Liquid |
Chemical Identifiers
7440-22-4 | |
107.87 | |
BQCADISMDOOEFD-UHFFFAOYSA-N | |
23954 | |
silver |
Ag | |
MFCD00003397 | |
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber | |
CHEBI:9141 | |
[Ag] |
Safety and Handling
TSCA : Yes
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only