Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver nanoparticles, 40nm, 0.02mg/mL, supplied in 2mM sodium citrate, 416nm absorption


Quantity 100mL

Manufacturer: alfa aesar™  J67090AE

 View more versions of this product

Catalog No. AAJ67090AE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

CAS 7440-22-4
Chemical Name or Material Silver nanoparticles
MDL Number MFCD00003397
TSCA TSCA
Name Note 0.02mg/mL, supplied in 2mM sodium citrate, 416nm absorption
Physical Form Liquid
Particle Size 40nm
Color Yellow
Storage Note 1 2° to 8°C
Odor Odorless
Quantity 100mL

Specifications

Chemical Name or Material Silver nanoparticles
CAS 7440-22-4
Name Note 0.02mg/mL, supplied in 2mM sodium citrate, 416nm absorption
MDL Number MFCD00003397
Accessories

2° to 8°C

SDS & Safety Documents