Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver nanoparticles, 30nm, 0.02mg/mL, supplied in 2mM sodium citrate, 410nm absorption


Quantity 25mL

Manufacturer: alfa aesar™  J67207AC

 View more versions of this product

Catalog No. AAJ67207AC

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Silver nanoparticles
MDL Number MFCD00003397
Name Note 0.02mg/mL, supplied in 2mM sodium citrate, 410nm absorption
Physical Form Liquid
CAS 7440-22-4
Particle Size 30nm
Sensitivity Light Sensitive
Storage Note 1 2° to 8°C
Color Yellow
Quantity 25mL

Specifications

Chemical Name or Material Silver nanoparticles
CAS 7440-22-4
Name Note 0.02mg/mL, supplied in 2mM sodium citrate, 410nm absorption
MDL Number MFCD00003397
Accessories

2° to 8°C