Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver nanoparticles, 100nm, 0.02mg/mL, supplied in 2mM sodium citrate, 490nm absorption


Quantity 100mL

Manufacturer: alfa aesar™  J67099AE

 View more versions of this product

Catalog No. AAJ67099AE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

CAS 7440-22-4
TSCA TSCA
Name Note 0.02mg/mL, supplied in 2mM sodium citrate, 490nm absorption
Quantity 100mL
Particle Size 100nm
MDL Number MFCD00003397
Physical Form Liquid
Color Yellow
Odor Odorless
Storage Note 1 2° to 8°C
Chemical Name or Material Silver nanoparticles

Specifications

Chemical Name or Material Silver nanoparticles
CAS 7440-22-4
Name Note 0.02mg/mL, supplied in 2mM sodium citrate, 490nm absorption
MDL Number MFCD00003397

  • An alternative to gold nanoparticles due to lower absorption maximum in the 400-500nm range
  • Readily binds proteins and other ligands for conjugate and sensor development

Accessories

2° to 8°C

SDS & Safety Documents