missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silver nanoparticles, 100nm, 0.02 mg/ml, supplied in 2mM sodium citrate, 490nm absorption, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J67099AC
Description
An alternative to gold nanoparticles due to lower absorption maximum in the 400-500nm range. Readily binds proteins and other ligands for conjugate and sensor development.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- An alternative to gold nanoparticles due to lower absorption maximum in the 400-500nm range
- Readily binds proteins and other ligands for conjugate and sensor development
Specifications
Yellow | |
0.02mg/mL, supplied in 2mM sodium citrate, 490nm absorption | |
Ag | |
25 mL | |
BQCADISMDOOEFD-UHFFFAOYSA-N | |
silver | |
23954 | |
Odorless | |
Light sensitive |
Silver nanoparticles | |
7440-22-4 | |
MFCD00003397 | |
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber | |
[Ag] | |
107.87 | |
CHEBI:9141 | |
100nm | |
Liquid |
Chemical Identifiers
7440-22-4 | |
107.87 | |
BQCADISMDOOEFD-UHFFFAOYSA-N | |
23954 | |
silver |
Ag | |
MFCD00003397 | |
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber | |
CHEBI:9141 | |
[Ag] |
Safety and Handling
TSCA : Yes
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only