missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver methanesulfonate, Alfa Aesar™

Manufacturer:  Alfa Aesar 1864109

 View more versions of this product

Catalog No. AA1864109



Specifications

Specifications

Silver methanesulfonate
CH3AgO3S
White powder
MLKQJVFHEUORBO-UHFFFAOYSA-M
silver;methanesulfonate
159658
Powder
White
2386-52-9
MFCD00064795
silver methanesulfonate, silver methylsulfonate, silver methanesulphonate, methanesulfonic acid, silver salt, silver methanesulfonate, silver salt, silver i methanesulfonate, methanesulfonic acid, silver 1+ salt, silver methanesulfonate ag ch2so3, methylsulfonic acid silver salt, silvermesylate
CS(=O)(=O)[O-].[Ag+]
202.96
202.97
Odorless
10g

Chemical Identifiers

2386-52-9
202.96
MLKQJVFHEUORBO-UHFFFAOYSA-M
159658
CS(=O)(=O)[O-].[Ag+]
CH3AgO3S
MFCD00064795
silver methanesulfonate, silver methylsulfonate, silver methanesulphonate, methanesulfonic acid, silver salt, silver methanesulfonate, silver salt, silver i methanesulfonate, methanesulfonic acid, silver 1+ salt, silver methanesulfonate ag ch2so3, methylsulfonic acid silver salt, silvermesylate
silver;methanesulfonate
Safety and Handling

Safety and Handling

H318-H315-H400-H335: Causes serious eye damage. Causes skin irritation. Very toxic to aquatic life. May cause respiratory irritation.

DOTInformation : Hazard Class: 9, Packing Group: III

EINECSNumber : 219-199-2

SDS Documents