missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver metavanadate

Manufacturer:  Alfa Aesar™ 4482806

 View more versions of this product

Catalog No. AA4482806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Silver metavanadate
AgO3V
MFCD00041760
silver metavanadate, silver vanadium trioxide, silver vanadium oxide agvo3, silver orthovanadate, silver vanadate v, silver metavanadate 5g, ag.o3v, silver oxido dioxo vanadium, silver vanadium oxide agvo3, silver 1+ dioxidovanadium 1+
[O-][V](=O)=O.[Ag+]
206.807
206.81
Odorless
Red
13497-94-4
AgVO3
UN3285
RAVDHKVWJUPFPT-UHFFFAOYSA-N
silver;oxido(dioxo)vanadium
3084155
Powder
Light Sensitive
5g

Chemical Identifiers

13497-94-4
206.807
RAVDHKVWJUPFPT-UHFFFAOYSA-N
3084155
[O-][V](=O)=O.[Ag+]
AgO3V
MFCD00041760
silver metavanadate, silver vanadium trioxide, silver vanadium oxide agvo3, silver orthovanadate, silver vanadate v, silver metavanadate 5g, ag.o3v, silver oxido dioxo vanadium, silver vanadium oxide agvo3, silver 1+ dioxidovanadium 1+
silver;oxido(dioxo)vanadium
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.

TSCA : TSCA

SDS Documents