missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver lactate

Manufacturer:  Alfa Aesar™ 2083518

 View more versions of this product

Catalog No. AA2083518



Specifications

Specifications

Silver lactate
C3H5AgO3
MFCD00043279
14,8517
LMEWRZSPCQHBOB-UHFFFAOYSA-M
silver;2-hydroxypropanoate
91824762
Powder
Light Sensitive
120°C to 122°C
128-00-7
CH3CH(OH)CO2Ag
UN3077
silver lactate, silver lactate monohydrate, lactato-o1,o2 silver, lactic acid, silver salt, silver, 2-hydroxypropanoato-o1,o2, propanoic acid, 2-hydroxy-, silver complex, silver, 2-hydroxy-kappao propanoato-kappao
CC(C(=O)[O-])O.[Ag+]
196.938
196.94
Odorless
White to Gray
50g

Chemical Identifiers

128-00-7
196.938
LMEWRZSPCQHBOB-UHFFFAOYSA-M
91824762
CC(C(=O)[O-])O.[Ag+]
C3H5AgO3
MFCD00043279
silver lactate, silver lactate monohydrate, lactato-o1,o2 silver, lactic acid, silver salt, silver, 2-hydroxypropanoato-o1,o2, propanoic acid, 2-hydroxy-, silver complex, silver, 2-hydroxy-kappao propanoato-kappao
silver;2-hydroxypropanoate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315
Harmful if swallowed.
Causes skin irritation.

GHS P Statement
P280g-P273-P305+P351+P338
Wear protective gloves.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : UA2465550

SDS Documents