Learn More
Silver iodide, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01211114
Description
Silver iodide is used in photographic film. Used in medical devices as a sustained release antibacterial. It also used in as an antiseptic and in cloud seeding. It is used in proteomics research applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver iodide | |
7783-96-2 | |
Powder | |
(metals basis) | |
MFCD00003412 | |
14,8516 | |
Soluble in alkali cyanides and iodides. Insoluble in water,acids and ammonium carbonate. | |
[Ag]I | |
234.773 | |
234.77 | |
Premion™ | |
Light sensitive |
558°C | |
Yellow | |
25 g | |
AgI | |
UN3077 | |
silver iodide, neosiluol, neosilvol, silver monoiodide, silver 1+ iodide, colloidal silver iodide, iodanylsilver, silver iodide metals basis, ccris 8490 | |
MSFPLIAKTHOCQP-UHFFFAOYSA-M | |
iodosilver | |
24563 | |
99.999% | |
Odorless | |
5.67 g/mL |
Chemical Identifiers
7783-96-2 | |
234.773 | |
MSFPLIAKTHOCQP-UHFFFAOYSA-M | |
24563 | |
[Ag]I |
AgI | |
MFCD00003412 | |
silver iodide, neosiluol, neosilvol, silver monoiodide, silver 1+ iodide, colloidal silver iodide, iodanylsilver, silver iodide metals basis, ccris 8490 | |
iodosilver |
Safety and Handling
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 232-038-0
RTECSNumber : VW4450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only