missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver iodide, 99.9% (metals basis) , Alfa Aesar™

Manufacturer:  Alfa Aesar 1141922

 View more versions of this product

Catalog No. AA1141922



Specifications

Specifications

Silver iodide
99.9% (metals basis)
MFCD00003412
MSFPLIAKTHOCQP-UHFFFAOYSA-M
iodosilver
24563
Crystalline
100g
7783-96-2
AgI
silver iodide, neosiluol, silver iodide, neosilvol, silver monoiodide, silver 1+ iodide, colloidal silver iodide, iodanylsilver, silver iodide metals basis, ccris 8490
[Ag]I
234.773
234.77
5.67g/mL

Chemical Identifiers

7783-96-2
234.773
MSFPLIAKTHOCQP-UHFFFAOYSA-M
24563
[Ag]I
AgI
MFCD00003412
silver iodide, neosiluol, silver iodide, neosilvol, silver monoiodide, silver 1+ iodide, colloidal silver iodide, iodanylsilver, silver iodide metals basis, ccris 8490
iodosilver
Safety and Handling

Safety and Handling

EINECSNumber : 232-038-0

Documents