Learn More
Silver(II) oxide, 98%, Thermo Scientific Chemicals
Manufacturing of silver oxide-zinc alkali batteries
Supplier: Thermo Scientific Chemicals 02269514
Description
Silver(II) oxide is used to make silver oxide-zinc alkali batteries. It is also used as a reagent in laboratory reactions and forms transition metal-carbene complexes. It is also used as a catalyst to prepare silver metal. It acts as an effective carbon dioxide scrubber and used in nuclear submarines, the international space station and space shuttles. Further, it is used in the manufacture of filters for gas sensors and in some infection-resistant surgical fabric materials and fibrous textile articles. In addition to this, it is used in concrete and in some swimming pools to protect the water from undesired microbes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver(II) oxide | |
1301-96-8 | |
Powder | |
AgO | |
UN3085 | |
silver ii oxide, silver oxide, silberperoxyd, silver oxide,ago, acmc-1aiyc, silver i,iii oxide, ksc203c6r, silver ii oxide 5g, silver ii oxide-ag | |
OTCVAHKKMMUFAY-UHFFFAOYSA-N | |
oxosilver | |
92152 | |
98% | |
Light and moisture sensitive |
>100°C (decomposition) | |
Black to Gray | |
25 g | |
MFCD00044285 | |
14,8522 | |
Insoluble in water; Soluble in alkalies,ammonium hydroxide with decomposition and N2 formation. | |
O=[Ag] | |
123.867 | |
123.87 | |
Odorless | |
7.44 g/mL |
Chemical Identifiers
1301-96-8 | |
123.867 | |
OTCVAHKKMMUFAY-UHFFFAOYSA-N | |
92152 | |
O=[Ag] |
AgO | |
MFCD00044285 | |
silver ii oxide, silver oxide, silberperoxyd, silver oxide,ago, acmc-1aiyc, silver i,iii oxide, ksc203c6r, silver ii oxide 5g, silver ii oxide-ag | |
oxosilver |
Safety and Handling
GHS H Statement
H272-H314-H318
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P370+P380+P375-P372-P380-P501c
H272-H314-H318-H335
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, CORROSIVE, N.O.S.
EINECSNumber : 215-098-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only