Please login to your online account to display your discounted pricing

Silver ICP Standard, 1mL = 10mg Ag (10,000ppm Ag), Ricca Chemical

Chemical Identifiers

Chemical Name or Material Silver ICP Standard
CAS 7732-18-5
CAS Min % 90.0
CAS Max % 94.0
CAS 7697-37-2
CAS Min % 6.75
CAS Max % 7.03
CAS 7761-88-8
CAS Min % 1.42
CAS Max % 1.47
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PAG10KN-100

 
ricca chemical company
RIPAG10KN100
Amber Poly 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 100mL
Chemical Name or Material Silver ICP Standard
CAS 7732-18-5
CAS Min % 90.0
CAS Max % 7.03
Concentration or Composition Notes 10,000ppm Ag
CAS 7761-88-8
Equivalence 1 mL = 10 mg Ag
CAS Max % 1.47
CAS Max % 94.0
CAS 7697-37-2
Packaging Amber Poly
CAS Min % 6.75
CAS Min % 1.42