Please login to your online account to display your discounted pricing

Silver(I) fluoride, 99+%, ACROS Organics™


Quantity 25g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  210450250

 View more versions of this product

Catalog No. AC210450250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Powder
Formula Weight 126.87
Quantity 25g
Color Brown-yellow to rust brown
Merck Index 15, 8653
EINECS Number 231-895-8
TSCA TSCA
RTECS Number VW4250000
Density 5.8500g/mL
Fieser 02,364; 05,581; 06,514; 15,287
Chemical Name or Material Silver(I) fluoride
Name Note 99+%
Packaging Plastic bottle
Assay Percent Range 99+%
Molecular Formula AgF
Linear Formula AgF
MDL Number MFCD00003410
CAS 7775-41-9
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Silver(I) fluoride
Name Note 99+%
CAS 7775-41-9
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula AgF
Linear Formula AgF
Formula Weight 126.87
MDL Number MFCD00003410
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents