Learn More
Silver(I) citrate hydrate, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04549622
Description
Silver(I) citrate hydrate can be very attractive for the future biomedical or pharmaceutical/therapeutic applications. It can be used as an antimicrobial agent and enhancers of antioxidants and anticancer agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver(I) citrate hydrate | |
170°C (decomposition) | |
MFCD00150589 | |
Light sensitive | |
Soluble in water. | |
[Ag+].[Ag+].[Ag+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O | |
512.70 | |
512.70 (Anhydrous) |
206986-90-5 | |
C6H5Ag3O7 | |
100 g | |
silver i citrate hydrate, trisilver 1+ citrate hydrate | |
QUTYHQJYVDNJJA-UHFFFAOYSA-K | |
2-hydroxypropane-1,2,3-tricarboxylic acid;silver;hydrate | |
16212313 | |
Powder |
Chemical Identifiers
206986-90-5 | |
512.70 | |
QUTYHQJYVDNJJA-UHFFFAOYSA-K | |
16212313 | |
[Ag+].[Ag+].[Ag+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O |
C6H5Ag3O7 | |
MFCD00150589 | |
silver i citrate hydrate, trisilver 1+ citrate hydrate | |
2-hydroxypropane-1,2,3-tricarboxylic acid;silver;hydrate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 628-014-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only