missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silver(I) behenate, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04549406
Description
Silver(I) behenate is used as a calibration standard in X-ray spectrophotometer to an absolute X-ray energy scale. It is utilized for low-angle diffraction standard in X-ray crystallography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver(I) behenate | |
White | |
Powder | |
C22H43AgO2 | |
silver behenate, silver docosanoate, behenic acid silver salt, unii-n3hp26f3pb, n3hp26f3pb, silver hebenate, docosanoic acid, silver 1+ salt 1:1, docosanoic acid silver salt, docosanoic acid, silver salt, docosanoic acid, silver 1+ salt | |
AQRYNYUOKMNDDV-UHFFFAOYSA-M | |
silver;docosanoate | |
164971 | |
Light sensitive |
(decomposition) | |
2489-05-6 | |
5 g | |
MFCD00059001 | |
Slightly soluble in water. | |
CCCCCCCCCCCCCCCCCCCCCC(=O)[O-].[Ag+] | |
447.45 | |
447.44 |
Chemical Identifiers
2489-05-6 | |
MFCD00059001 |
Safety and Handling
EINECSNumber : 219-641-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only