missing translation for 'onlineSavingsMsg'
Learn More

Silver hexafluorophosphate, 98%

CAS: 26042-63-7 | AgF6P | 252.832 g/mol

Supplier:  Thermo Scientific Chemicals 01187306

 Afficher plus de versions de ce produit

Numéro de catalogue. AA1187306


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

It is commonly used as an oxidant forming silver metal. It is often used as a reagent in inorganic and organometallic chemistry. It is used in the preparation of engineering novel materials as well as in preparation of supra molecular Ag(I) complexes. It is used in the development of new catalysts, replacing halide ligands with the weakly coordinating hexafluorophosphate anion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

Silver hexafluorophosphate
26042-63-7
Powder
98%
MFCD00003415
Soluble in organic solvents such as benzene,toluene,and acetonitrile.
F[P-](F)(F)(F)(F)F.[Ag+]
252.832
252.83
Light sensitive
102°C (decomposition)
Brown to Yellow
5 g
AgF6P
silver hexafluorophosphate, silver hexafluorophosphate v, ag.f6p, silver i hexafluorophosphate, agpf6, acmc-1ceef, phosphate 1-, hexafluoro-, silver 1+, phosphate 1-, hexafluoro-, silver 1+ 1:1, silver i hexafluorophosphate v
SCQBROMTFBBDHF-UHFFFAOYSA-N
silver;hexafluorophosphate
168464
Odorless
White powder

Identifiants chimiques

26042-63-7
252.832
SCQBROMTFBBDHF-UHFFFAOYSA-N
168464
F[P-](F)(F)(F)(F)F.[Ag+]
AgF6P
MFCD00003415
silver hexafluorophosphate, silver hexafluorophosphate v, ag.f6p, silver i hexafluorophosphate, agpf6, acmc-1ceef, phosphate 1-, hexafluoro-, silver 1+, phosphate 1-, hexafluoro-, silver 1+ 1:1, silver i hexafluorophosphate v
silver;hexafluorophosphate
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

missing translation for 'dotInformation' : DOT Class: 8; Packaging Group: II

missing translation for 'einecsNumber' : 247-428-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only