Learn More
Silver hexafluorophosphate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01187314
Description
It is commonly used as an oxidant forming silver metal. It is often used as a reagent in inorganic and organometallic chemistry. It is used in the preparation of engineering novel materials as well as in preparation of supra molecular Ag(I) complexes. It is used in the development of new catalysts, replacing halide ligands with the weakly coordinating hexafluorophosphate anion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver hexafluorophosphate | |
26042-63-7 | |
Powder | |
98% | |
MFCD00003415 | |
Soluble in organic solvents such as benzene,toluene,and acetonitrile. | |
F[P-](F)(F)(F)(F)F.[Ag+] | |
252.832 | |
252.83 | |
Light sensitive |
102°C (decomposition) | |
Brown to Yellow | |
25 g | |
AgF6P | |
silver hexafluorophosphate, silver hexafluorophosphate v, ag.f6p, silver i hexafluorophosphate, agpf6, acmc-1ceef, phosphate 1-, hexafluoro-, silver 1+, phosphate 1-, hexafluoro-, silver 1+ 1:1, silver i hexafluorophosphate v | |
SCQBROMTFBBDHF-UHFFFAOYSA-N | |
silver;hexafluorophosphate | |
168464 | |
Odorless | |
White powder |
Chemical Identifiers
26042-63-7 | |
252.832 | |
SCQBROMTFBBDHF-UHFFFAOYSA-N | |
168464 | |
F[P-](F)(F)(F)(F)F.[Ag+] |
AgF6P | |
MFCD00003415 | |
silver hexafluorophosphate, silver hexafluorophosphate v, ag.f6p, silver i hexafluorophosphate, agpf6, acmc-1ceef, phosphate 1-, hexafluoro-, silver 1+, phosphate 1-, hexafluoro-, silver 1+ 1:1, silver i hexafluorophosphate v | |
silver;hexafluorophosphate |
Safety and Handling
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : DOT Class: 8; Packaging Group: II
EINECSNumber : 247-428-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only