missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver hexafluoroantimonate, 99%

Manufacturer:  Alfa Aesar™ 1153814

 View more versions of this product

Catalog No. AA1153814



Specifications

Specifications

Silver hexafluoroantimonate
AgF6Sb
UN3260
GSXFODUDOAXUBF-UHFFFAOYSA-H
silver;hexafluoroantimony(1-)
16687962
Powder/Lump
Light Sensitive & Hygroscopic
25g
26042-64-8
MFCD00003401
acmc-209tq8
F[Sb-](F)(F)(F)(F)F.[Ag+]
343.619
343.41
99%
White to Cream

Chemical Identifiers

26042-64-8
343.619
GSXFODUDOAXUBF-UHFFFAOYSA-H
16687962
F[Sb-](F)(F)(F)(F)F.[Ag+]
AgF6Sb
MFCD00003401
acmc-209tq8
silver;hexafluoroantimony(1-)
Safety and Handling

Safety and Handling

GHS H Statement
H314-H302-H332
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

Documents