Learn More
Silver hexafluoroantimonate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 312500250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Silver hexafluoroantimonate | |
25g | |
AgF6Sb | |
MFCD00003401 | |
silver hexafluoroantimonate, silver hexafluoroantimonate v, silver hexafluorostiboranuide, silver 1+ hexafluorostibanuide, ag.f6sb, silverhexafluoroantimonate, agsbf6, acmc-209tq8, ksc492g5j, silver i hexafluoroantimonate | |
GSXFODUDOAXUBF-UHFFFAOYSA-H | |
silver;hexafluoroantimony(1-) | |
16687962 | |
98% |
26042-64-8 | |
97.5% min. (Argentometry) | |
AgSbF6 | |
05,577; 07,320; 17,17 | |
Solubility in water: soluble | |
F[Sb-](F)(F)(F)(F)F.[Ag+] | |
343.61 | |
343.61 | |
Plastic bottle |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
GHS Signal Word: Danger
EINECSNumber : 247-429-1
RUO â Research Use Only