missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver hexafluoroantimonate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 312500050

 View more versions of this product

Catalog No. AC312500050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Silver hexafluoroantimonate
Crystalline Powder
98%
AgSbF6
MFCD00003401
Plastic bottle
5g
26042-64-8
≥97.5%
AgF6Sb
343.61
05,577; 07,320; 17,17
White to pale yellow

Chemical Identifiers

98%
AgSbF6
MFCD00003401
≥97.5%
AgF6Sb
343.61
Silver hexafluoroantimonate

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger