missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver/Gray Fluormag™ Magnetic Latent Print Powder

Highly fluorescent for use on multicolored backgrounds.

$35.40 - $35.40

Specifications

Color Silver Gray
For Use With (Application) For use on multicolored backgrounds
View More Specs

Products 1
Catalog Number Mfr. No. Product Type Weight (English) Weight (Metric) Price Quantity  
Catalog Number Mfr. No. Product Type Weight (English) Weight (Metric) Price Quantity  
S99533
View Documents
Fisher Scientific
FMP01
Magnetic Latent Print Powder 2 oz. 56g
Each for $35.40
 
Description

Description

  • “No contact” magnetic powder
  • Natural silver/gray color appears bright yellow/green when illuminated with an ultraviolet light

Specifications

Specifications

Silver Gray
For use on multicolored backgrounds
SDS