Silver/Gray Fluormag™ Magnetic Latent Print Powder
Highly fluorescent for use on multicolored backgrounds.
Manufacturer:  Fisher Scientific FMP0116
 View more versions of this product
Catalog No. S69013D
Description
Description
  • “No contact” magnetic powder
  • Natural silver/gray color appears bright yellow/green when illuminated with an ultraviolet light
Specifications
Specifications
Silver Gray
Magnetic Print Powder
453.5g
For use on multicolored backgrounds
16 oz.
Education
Documents