missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver gauze, 20 mesh woven from 0.356mm (0.014 in.) dia. wire

Filtration, electrodes, contacts

Manufacturer:  Alfa Aesar™ 44449FL

 View more versions of this product

Catalog No. AA44449FL



Specifications

Specifications

Silver gauze
20 mesh woven from 0.356mm (0.014 in.) dia. wire
Ag
20 Mesh
Insoluble in water.
[Ag]
Characteristic
23954
Wire Cloth
75 x 75mm
7440-22-4
MFCD00003397
algaedyn
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
107.868
CHEBI:9141

Chemical Identifiers

7440-22-4
107.868
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
algaedyn
CHEBI:9141
[Ag]
Safety and Handling

Safety and Handling

EINECSNumber : 231-131-3

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents