missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silver gauze, 20 mesh woven from 0.356mm (0.014in) dia wire, Thermo Scientific Chemicals
Filtration, electrodes, contacts
Supplier: Thermo Scientific Chemicals 044449HB
Description
Silver gauze is used for filtration. It is also used in electrodes and contacts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver gauze | |
7440-22-4 | |
MFCD00003397 | |
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber | |
BQCADISMDOOEFD-UHFFFAOYSA-N | |
silver | |
23954 | |
20 Mesh | |
Wire Cloth |
20 mesh woven from 0.356mm (0.014 in.) dia. wire | |
Ag | |
300 x 300 mm | |
Insoluble in water. | |
[Ag] | |
107.87 | |
CHEBI:9141 | |
Characteristic |
Chemical Identifiers
7440-22-4 | |
107.87 | |
BQCADISMDOOEFD-UHFFFAOYSA-N | |
23954 | |
silver |
Ag | |
MFCD00003397 | |
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber | |
CHEBI:9141 | |
[Ag] |
Safety and Handling
EINECSNumber : 231-131-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only