missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver Foil, 2.0mm (0.08 in.) thick, Premion™, 99.9985% (Metals basis), Alfa Aesar™

Form: Approx. 13g/25 x 25mm

Manufacturer:  Thermo Fisher Scientific 12129FF

 View more versions of this product

Catalog No. AA12129FF



Specifications

Specifications

Silver
7440-22-4
99.9985% (Metals basis)
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
107.868
CHEBI:9141
Silver
25 x 25mm
Silver-colored foil
Ag
algaedyn, amalgum, argentum, atom, colloidal, epinall, metal, silber, silver, colloidal, silver, elemental
[Ag]
Characteristic
23954
Foil, 2.0mm (0.08 in.) thick

Chemical Identifiers

7440-22-4
107.868
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
algaedyn, amalgum, argentum, atom, colloidal, epinall, metal, silber, silver, colloidal, silver, elemental
CHEBI:9141
[Ag]
Safety and Handling

Safety and Handling

EINECSNumber : 231-131-3

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents