Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver foil, 2.0mm (0.08 in.) thick, hard, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Silver foil
CAS 7440-22-4
Name Note 2.0mm (0.08 in.) thick, Hard, Temper: spring hard
MDL Number MFCD00003397
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA11437FF

 
alfa aesar™
11437FF
25x25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA11437FI

 
alfa aesar™
11437FI
50x50mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA11437GH

 
alfa aesar™
11437GH
100x100mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Silver foil
TSCA TSCA
Name Note 2.0mm (0.08 in.) thick, Hard, Temper: spring hard
Physical Form Foil
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00003397
CAS 7440-22-4
Odor Characteristic
Weight ≈13g/25x25mm
Quantity 25x25mm