missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver foil, 1.0mm (0.04 in.) thick, hard, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 11438FI

 View more versions of this product

Catalog No. AA11438FI



Specifications

Specifications

Silver foil
50 x 50mm
7440-22-4
Ag
algaedyn
[Ag]
Characteristic
23954
Foil
≈6.6g/25x25mm
1.0mm (0.04 in.) thick, Hard, Temper: spring hard
(metals basis)
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
107.868
CHEBI:9141
99.9%

Chemical Identifiers

7440-22-4
107.868
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
algaedyn
CHEBI:9141
[Ag]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents