missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver Foil, 0.25mm (0.01 in.) thick, hard, Premion™, 99.998% (Metals basis), Alfa Aesar™

Form: Approx. 6.6g/50 x 50mm; Temper: as rolled

Manufacturer:  Alfa Aesar 12125GH

 View more versions of this product

Catalog No. AA12125GH



Specifications

Specifications

Silver
Hard
Silver-colored foil
Ag
algaedyn
[Ag]
Characteristic
23954
Foil, 0.25mm (0.01 in.) thick
100 x 100mm
7440-22-4
99.998% (Metals basis)
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
107.868
CHEBI:9141
Silver

Chemical Identifiers

7440-22-4
107.868
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
algaedyn
CHEBI:9141
[Ag]
Documents