missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver Foil, 0.1mm (0.004 in.) thick, hard, Premion™, 99.998% (Metals basis), Alfa Aesar™

Form: Approx. 2.6g/50 x 50mm; Temper: as rolled

Manufacturer:  Alfa Aesar 12126GH

 View more versions of this product

Catalog No. AA12126GH



Specifications

Specifications

Silver
Hard
Silver-colored foil
Ag
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
[Ag]
Characteristic
23954
Foil, 0.1mm (0.004 in.) thick
100 x 100mm
7440-22-4
99.998% (Metals basis)
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
107.868
CHEBI:9141
Silver

Chemical Identifiers

7440-22-4
107.868
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
CHEBI:9141
[Ag]
Documents