missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver Foil, 0.025mm (0.001 in.) thick, annealed, 99.95% (Metals basis), Alfa Aesar™

Form: Approx. 5.9g/150 x 150mm; Temper: soft

Manufacturer:  Alfa Aesar 121901EX

 View more versions of this product

Catalog No. AA121901EX



Specifications

Specifications

Silver foil
7440-22-4
Ag
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
[Ag]
Characteristic
23954
≈5.9g/150X150mm, Temper: Soft
0.025mm (0.001 in.) thick, annealed
(metals basis)
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
107.868
CHEBI:9141
99.95%

Chemical Identifiers

7440-22-4
107.868
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
CHEBI:9141
[Ag]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents