Please login to your online account to display your discounted pricing

Silver foil, 0.025mm (0.001in) thick, annealed, 99.95% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Silver foil
CAS 7440-22-4
Name Note 0.025mm (0.001 in.) thick, annealed
MDL Number MFCD00003397
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

AA121901EX

  SDS
alfa aesar
121901EX
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form ≈5.9g/150X150mm, Temper: Soft
TSCA TSCA
CAS 7440-22-4
Odor Characteristic
Chemical Name or Material Silver foil
Name Note 0.025mm (0.001 in.) thick, annealed
Assay Percent Notes (metals basis)
MDL Number MFCD00003397
Assay 99.95%