missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver flake, 90% <20 micron, 99.9% (metals basis)

Suitable for manufacturing metallizing preparations and conductive silicone rubbers

Manufacturer:  Alfa Aesar™ 1475509

 View more versions of this product

Catalog No. AA1475509



Specifications

Specifications

10g
7440-22-4
(metals basis)
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
[Ag]
107.868
CHEBI:9141
Flakes
Silver flake
99.9%
Ag
UN3077
Characteristic
silver
23954
90% <20 Micron

Chemical Identifiers

7440-22-4
107.868
BQCADISMDOOEFD-UHFFFAOYSA-N
CHEBI:9141
[Ag]
Ag
MFCD00003397
23954
silver
Safety and Handling

Safety and Handling

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

TSCA : TSCA

SDS Documents