missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver flake, 90% <20 micron, 99.9% (metals basis), Thermo Scientific Chemicals

Suitable for manufacturing metallizing preparations and conductive silicone rubbers

Supplier:  Thermo Scientific Chemicals 01475550

 View more versions of this product

Catalog No. AA1475550



Description

Description

Suitable for manufacturing metallizing preparations and conductive silicone rubbers

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Silver flake
(metals basis)
MFCD00003397
5 x 10 g
BQCADISMDOOEFD-UHFFFAOYSA-N
silver
23954
99.9%
90% <20 Micron
7440-22-4
Ag
UN3077
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
[Ag]
107.87
CHEBI:9141
Characteristic
Flake

Chemical Identifiers

7440-22-4
107.87
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
CHEBI:9141
[Ag]
Safety and Handling

Safety and Handling

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 231-131-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only