missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver flake, 80% <20 micron, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 1463930

 View more versions of this product

Catalog No. AA1463930



Specifications

Specifications

Silver flake
250g
(metals basis)
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
Characteristic
silver
23954
80% <20 Micron
99.9%
Scott density: 18g/cm3 to 28g/in3
7440-22-4
Ag
UN3077
BQCADISMDOOEFD-UHFFFAOYSA-N
[Ag]
107.868
CHEBI:9141
Flakes
1.0 to 2.2m2/g

Chemical Identifiers

7440-22-4
107.868
BQCADISMDOOEFD-UHFFFAOYSA-N
23954
silver
Ag
MFCD00003397
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
CHEBI:9141
[Ag]
Safety and Handling

Safety and Handling

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

TSCA : TSCA

SDS Documents