Please login to your online account to display your discounted pricing

Silver Diethyldithiocarbamate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 10g
Packaging Wide Mouth Amber Glass Bottle

Manufacturer: avantor performance  H73902

 View more versions of this product

Catalog No. 02003923

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 256.14
CAS 1470-61-7
Chemical Name or Material Silver Diethyldithiocarbamate
Molecular Formula (C2H5)2NCSSAg
Packaging Wide Mouth Amber Glass Bottle
Quantity 10g

Specifications

Chemical Name or Material Silver Diethyldithiocarbamate
CAS 1470-61-7
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula (C2H5)2NCSSAg
Formula Weight 256.14