Please login to your online account to display your discounted pricing

Silver Diethyldithiocarbamate, ACS Grade, LabChem™

$642.86 - $642.86

Chemical Identifiers

Chemical Name or Material Silver Diethyldithiocarbamate
CAS 1470-61-7
CAS Max % 100
Purity Grade ACS Grade
Linear Formula C5H10AgNS2
Formula Weight 256.14
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC224258

 
lab chem
LC224258
25g Each for $642.86
Description & Specifications

Specifications

Formula Weight 256.14
Health Hazard 1 Warning
Purity Grade ACS Grade
Health Hazard 3 GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Chemical Name or Material Silver Diethyldithiocarbamate
Identification Passes Test
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
EINECS Number 216-003-7
Decomposition Information Sulfur compounds; Carbon monoxide; Carbon dioxide; Nitrogen oxides; Silver
Quantity 25g
CAS Max % 100
Physical Form Powder
CAS 1470-61-7
Suitability Passes Test
Packaging Amber Glass
Linear Formula C5H10AgNS2
Color Slightly yellow
Storage Information Refrigerate
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.