Please login to your online account to display your discounted pricing

Silver Diethyldithiocarbamate, 0.3% in Chloroform, for Arsenic, LabChem™

$229.37 - $229.37

Chemical Identifiers

Chemical Name or Material Silver Diethyldithiocarbamate
CAS 67-66-3
CAS Max % 97.713
CAS 110-91-8
CAS Max % 1
CAS 64-17-5
CAS Max % 0.987
CAS 1470-61-7
CAS Max % 0.3
Name Note 0.3% in Chloroform, for Arsenic
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC224301

 
lab chem
LC224301
500mL Each for $229.37
Description & Specifications

Specifications

Formula Weight 256.14
Chemical Name or Material Silver Diethyldithiocarbamate
CAS Max % 0.987
Decomposition Information Chlorine; Carbon monoxide; Carbon dioxide; Sulfur compounds
CAS 1470-61-7
Color Clear, light yellow
Quantity 500mL
CAS Max % 0.3
Name Note 0.3% in Chloroform, for Arsenic
Physical Form Liquid
Linear Formula C5H10AgNS2
CAS 67-66-3
UN Number UN1888
Health Hazard 1 Danger
CAS Max % 97.713
Health Hazard 2 GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing cancer (inhalation).
Toxic to aquatic life.
Identification Passes Test
CAS 110-91-8
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Amber Glass
CAS Max % 1
CAS 64-17-5
Storage Information Refrigerate
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing cancer (inhalation).
Toxic to aquatic life.