missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver cyclohexanebutyrate, ACROS Organics™

Chemical Identifiers

Molecular Formula C10H17AgO2
Linear Formula C6H11(CH2)3COOAg
Formula Weight 277.1
MDL Number MFCD00036396
Chemical Name or Material Silver cyclohexanebutyrate
Assay 37.5 to 40.3% (Ag)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC419320010 SDS Thermo Fisher Scientific
AC419320010
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

C10H17AgO2
277.1
Silver cyclohexanebutyrate
C6H11(CH2)3COOAg
MFCD00036396
37.5 to 40.3% (Ag)

Specifications

62638-04-4
C10H17AgO2
C6H11(CH2)3COOAg
MFCD00036396
1g
37.5 to 40.3% (Ag)
Powder and/or Chunks
Authentic
277.1
Off-white to beige
Silver cyclohexanebutyrate

Safety and Handling

EINECSNumber : 263-667-9

TSCA : TSCA