Please login to your online account to display your discounted pricing

Silver cyclohexanebutyrate, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC419320010

 View more versions of this product

Catalog No. AC419320010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder and/or Chunks
Formula Weight 277.1
Quantity 1g
Color Off-white to beige
Assay 37.5 to 40.3% (Ag)
EINECS Number 263-667-9
TSCA TSCA
Infrared Spectrum Authentic
Chemical Name or Material Silver cyclohexanebutyrate
Molecular Formula C10H17AgO2
MDL Number MFCD00036396
Linear Formula C6H11(CH2)3COOAg
CAS 62638-04-4

Specifications

Chemical Name or Material Silver cyclohexanebutyrate
Assay 37.5 to 40.3% (Ag)
Molecular Formula C10H17AgO2
Linear Formula C6H11(CH2)3COOAg
Formula Weight 277.1
MDL Number MFCD00036396