missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver cyclohexanebutyrate, 99% (dry wt.)

$74.51 - $252.45

Chemical Identifiers

CAS 62638-04-4
Molecular Formula C10H17AgO2
Formula Weight 277.11
MDL Number MFCD00036396
Synonym Cyclohexanebutyric acid silver salt; 4-Cyclohexylbutyric acid silver salt
Chemical Name or Material Silver cyclohexanebutyrate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2261303 SDS Alfa Aesar™
B2261303
1g Each for $74.51
AAB2261306 SDS Alfa Aesar™
B2261306
5g Each for $252.45
Specifications Description & Specifications

Chemical Identifiers

62638-04-4
277.11
Cyclohexanebutyric acid silver salt; 4-Cyclohexylbutyric acid silver salt
C10H17AgO2
MFCD00036396
Silver cyclohexanebutyrate

Specifications

277.11
MFCD00036396
1g
62638-04-4
99%, (dry wt.)
C10H17AgO2
≈255°C (dec.)
Cyclohexanebutyric acid silver salt; 4-Cyclohexylbutyric acid silver salt
Silver cyclohexanebutyrate

Safety and Handling

TSCA : TSCA