Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver cyclohexanebutyrate

$20.49 - $71.57

Chemical Identifiers

Chemical Name or Material Silver cyclohexanebutyrate
CAS 62638-04-4
Molecular Formula C10H17AgO2
Formula Weight 277.11
MDL Number MFCD00036396
Synonym Cyclohexanebutyric acid silver salt; 4-Cyclohexylbutyric acid silver salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4370803

  SDS
alfa aesar™
4370803
1g Each for $20.49

AA4370806

  SDS
alfa aesar™
4370806
5g Each for $71.57
Description & Specifications

Specifications

Formula Weight 277.11
Synonym Cyclohexanebutyric acid silver salt; 4-Cyclohexylbutyric acid silver salt
Physical Form Powder
Quantity 1g
Sensitivity Light Sensitive
Molecular Formula C10H17AgO2
Chemical Name or Material Silver cyclohexanebutyrate
CAS 62638-04-4
TSCA TSCA
Melting Point ≈255°C (dec.)
MDL Number MFCD00036396