missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver cyanate, 98%, Alfa Aesar™

Manufacturer:  Alfa Aesar 4541114

 View more versions of this product

Catalog No. AA4541114



Specifications

Specifications

Silver cyanate
98%
MFCD00011512
VAQVDZZAAMAZAO-UHFFFAOYSA-N
cyanic acid;silver
45051745
Powder
White
3315-16-0
CHAgNO
silver,cyanate, cyanic acid; silver
C(#N)O.[Ag]
150.893
149.89
Odorless
25g

Chemical Identifiers

3315-16-0
150.893
VAQVDZZAAMAZAO-UHFFFAOYSA-N
45051745
C(#N)O.[Ag]
CHAgNO
MFCD00011512
silver,cyanate, cyanic acid; silver
cyanic acid;silver
Safety and Handling

Safety and Handling

H400-H302: Very toxic to aquatic life. Harmful if swallowed.

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 222-006-4

SDS Documents