Please login to your online account to display your discounted pricing

Silver cyanate, 98+%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  223710500

 View more versions of this product

Catalog No. AC223710500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Formula Weight 149.89
Physical Form Powder
Quantity 50g
Color Beige to gray
Chemical Name or Material Silver cyanate
Linear Formula AgOCN
CAS 3315-16-0
Assay Percent Range 98+%
Packaging Glass bottle
Molecular Formula CAgNO

Specifications

Chemical Name or Material Silver cyanate
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula CAgNO
Linear Formula AgOCN
Formula Weight 149.89