Please login to your online account to display your discounted pricing

Silver cyanate, 98+%, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC223712500

 View more versions of this product

Catalog No. AC223712500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Silver cyanate
CAS 3315-16-0
Physical Form Powder
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula CAgNO
Linear Formula AgOCN
Formula Weight 149.89
Packaging Glass bottle
Color Beige to gray
Quantity 250g

Specifications

Chemical Name or Material Silver cyanate
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula CAgNO
Linear Formula AgOCN
Formula Weight 149.89